Masterflex® L/S® Tang Adapter Kits, Avantor®
Supplier: Avantor Fluid Handling
7519-01
MFLX07519-01EA 73.1 USD
MFLX07519-01
Masterflex® L/S® Tang Adapter Kits, Avantor®
Pumps Peristaltic Pumps
Extend the life of your Masterflex pump head with these replacement parts.
  • Suitable for use with PTFE Diaphragm pump head and Multichannel Cartridge pump heads
Replacement parts to keep your pump head operating even longer. Tang adapter kits provides a tight tang shaft connection and reduces noise. Use large adapter for 75W (1/10 hp) drives and small adapter for 37W (1/20 hp drives).