Magnetic-Mount Liquid Level Float Switch
Supplier: VWR International
MFLX07187-15
MFLX07187-15EA 417 USD
MFLX07187-15
Magnetic-Mount Liquid Level Float Switch
Level Sensors
Permanent magnet attaches to steel tank wall at exact level required.
  • Constructed of brass, Buna N, aluminum, and 316 stainless steel
Use this normally open switch for filling operations and to prevent overflows in steel tanks. No need to drill mounting holes or provide piping — mount the switch at the exact level you need quickly and easily using the permanent magnet on the housing. Electrical connections are made with the 10-ft (3-m) neoprene-insulated cable.