Micropump Pump Head Adapter Kits
Supplier: MICROPUMP INC
81518 QPM 81518+213003
MFLX07003-99EA 600 USD
MFLX07003-99 MFLX07104-48 MFLX07003-96
Micropump Pump Head Adapter Kits
Pumps Gear Pumps
Easily adapt your pump head to your drive with these adapter kits.
  • Choose adapter kit suitable for use with your Masterflex pump head
Use these adapter kits to attach Micropump® A-Mount pump heads to your own motor or Masterflex® L/S® pump drives.