Flux® Heavy-Duty Drum Pump Motors, TEFC
Supplier: FLUX PUMPS CORP
F458-1 120V MFLX07076-25
MFLX07076-20EA 1840 USD
MFLX07076-20 MFLX07076-25
Flux® Heavy-Duty Drum Pump Motors, TEFC
Pumps Drum/Transfer Pumps
Configurable to your requirements.
  • High-viscosity tube set handles fluids with viscosities to 50,000 cp
Heavy-duty motors are best for applications requiring high output head. Motors have a built-in overload switch with manual rest to avoid the danger of automatic restarting.
Order tube set and accessories separately.