Gast® Direct-Drive Hub-Mount Air Motors
Supplier: GAST MANUFACTURING CORP
1UP-NRV-3A MFLX07055-72
MFLX07055-70EA 395 USD
MFLX07055-70 MFLX07055-72
Gast® Direct-Drive Hub-Mount Air Motors
Pumps Motors
Four-vane rotary design is almost maintenance free.
  • Nonsparking, nonelectrical operation—safe and reliable
These Gast® Direct-Drive Hub-Mount Air Motors feature a 1/2 hp reversible motor.
Delivery information: Includes motors come with an exhaust muffler. Install a compressed-air filter in-line to remove oil, water, and solid contaminants—order connecting nipples and filters separately.