Bev-A-Line® V Transfer Pump Tubing
Supplier: THERMOPLASTIC PROCESSES
B-5 B-9 B-13 B-19 B-38 MFLX06491-16
MFLX06491-12CS 0 USD
MFLX06491-12 MFLX06491-14 MFLX06491-15 MFLX06491-17 MFLX06491-39 MFLX06491-16
Bev-A-Line® V Transfer Pump Tubing
Tubing Transfer Tubing
Needs no connectors—heat-bondable.
  • Ideal for general-purpose, single-use, and extended fluid path applications
Applications: Safe for food, beverage, drug, lab, and general use—can withstand higher temperatures than Bev-A-Line® IV tubing. Safe for most alcohols; avoid strong solvents.
Characteristics: Polyethylene liner cross-linked to ethyl vinyl acetate shell. No plasticizers. Nontoxic and nonhemolytic. Needs no connectors—heat-bondable. Translucent.
Certification: Resin is FDA-compliant (liner: 21 CFR 177.1520; shell 21 CFR 177.1350). Meets USP Class VI, USDA, and National Formulary requirements.
Temperature range: –60 to 200 °F (–51 to 93 °C)
Sterilize: With autoclave, ethylene oxide, or dry heat.