Masterflex® L/S® High-Performance Precision Pump Tubing, Tygon® S3™ E-LFL, Avantor®
Supplier: VWR International
MFLX06440-15 MFLX06440-24 MFLX06440-35 MFLX06440-36
MFLX06440-15CS 0 USD
MFLX06440-15 MFLX06440-24 MFLX06440-35 MFLX06440-36
Masterflex® L/S® High-Performance Precision Pump Tubing, Tygon® S3™ E-LFL, Avantor®
Tubing Peristaltic Pump Tubing
Longest life of all Tygon® formulations.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
Applications: General laboratory use
Characteristics: Long life, won’t age or oxidize, fits tightly to glass and metal, bends to small radius curves, nontoxic, transparent
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.