Masterflex® Correlated Variable-Area Flowmeters, 150-mm Scale, Avantor®
Supplier: AALBORG INSTRUMENTS & CONTROL
P03/1-N032-41ST P03/1-N062-01ST P03/1-N112-02G P03/1-N082-03G P03/1-N112-02ST P03/1-N082-03ST P03/1-N092-04G P03/1-N102-05G P03/1-N102-05ST P03/1-N034-39G P03/1-N034-39ST T53/1-N062-01G T53/1-N112-02G T53/1-N112-02S T53/1-N092-04G T53/1-N034-39G P23/1-N032-41ST P23/1-N112-02ST P23/1-N082-03G P23/1-N112-02G P23/1-N082-03ST P23/1-N102-05G P23/1-N102-05ST P23/1-N034-39ST P13/1-N032-41G P13/1-N082-03ST MFLX03267-08 MFLX03267-22 MFLX03267-50 MFLX03267-67 MFLX03267-77 MFLX03267-80 MFLX03267-85 MFLX03269-03 MFLX03269-09 MFLX03269-16 MFLX03269-21 MFLX03269-23 MFLX03269-50 MFLX03269-62 MFLX03269-66 MFLX03269-70 MFLX03269-72 MFLX03269-74 MFLX03269-76 MFLX03269-78 MFLX03269-80
MFLX03267-00EA 201 USD
MFLX03267-00 MFLX03267-02 MFLX03267-06 MFLX03267-09 MFLX03267-15 MFLX03267-16 MFLX03267-17 MFLX03267-18 MFLX03267-20 MFLX03267-24 MFLX03267-26 MFLX03267-28 MFLX03267-30 MFLX03267-57 MFLX03267-62 MFLX03267-65 MFLX03267-72 MFLX03267-82 MFLX03269-07 MFLX03269-15 MFLX03269-17 MFLX03269-18 MFLX03269-19 MFLX03269-25 MFLX03269-27 MFLX03269-31 MFLX03269-52 MFLX03269-54 MFLX03269-68 MFLX03267-08 MFLX03267-22 MFLX03267-50 MFLX03267-67 MFLX03267-77 MFLX03267-80 MFLX03267-85 MFLX03269-03 MFLX03269-09 MFLX03269-16 MFLX03269-21 MFLX03269-23 MFLX03269-50 MFLX03269-62 MFLX03269-66 MFLX03269-70 MFLX03269-72 MFLX03269-74 MFLX03269-76 MFLX03269-78 MFLX03269-80
Masterflex® Correlated Variable-Area Flowmeters, 150-mm Scale, Avantor®
Flow Meters
Don’t sacrifice suitability for precision.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. PTFE has the best chemical compatibility.