Masterflex® Bag Filters for Polypropylene Bag Filter Housings, Avantor®
Supplier: PENTAIR RESIDENTIAL FILTRATION LLC
155383-03 155385-03 155391-03 155386-03 155396-03 255003-03 255024-03 255026-03 255028-03 255034-03 255037-03 MFLX01516-70 MFLX01516-76 MFLX01516-94 MFLX01519-02
MFLX01516-00EA 12 USD
MFLX01516-00 MFLX01516-02 MFLX01516-06 MFLX01516-08 MFLX01516-12 MFLX01516-20 MFLX01516-22 MFLX01516-24 MFLX01516-26 MFLX01516-28 MFLX01516-30 MFLX01516-32 MFLX01516-40 MFLX01516-44 MFLX01516-62 MFLX01516-72 MFLX01516-80 MFLX01516-84 MFLX01516-88 MFLX01519-06 MFLX01519-12 MFLX01519-16 MFLX01516-70 MFLX01516-76 MFLX01516-94 MFLX01519-02
Masterflex® Bag Filters for Polypropylene Bag Filter Housings, Avantor®
Filters Bag Filter Systems
Conveniently collect all filtered particles within the bag.
  • Choose from a wide range of bag materials suitable for your application
Choose from standard felt bags, high-efficiency bags, or nylon mesh bags. Each bag measures 4" diameter. The 10"L bags have a surface area of 0.75 ft2; 18-½"L bags have a surface area of 1.75 ft2.