Suc-Tyr-Val-Ala-Asp-Pna

Supplier: BACHEM AMERICAS INC

4017745.0025
L-1830.0025BAEA 334.97 USD
L-1830.0025BA
Suc-Tyr-Val-Ala-Asp-Pna
Suc-Tyr-Val-Ala-Asp-Pna

Sequence: Suc-Tyr-Val-Ala-Asp-pNA
Synonym(s): Suc-YVAD-pNA


Suc-YVAD-pNA, a chromogenic caspase-1 substrate.

Formula: C₃₁H₃₈N₆O₁₂
Storage Temperature: Freezer
MDL Number: MFCD01318836
CAS Number: 208264-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR