H-D-Val-Leu-Lys-pNA · 2 HCl
Supplier: BACHEM AMERICAS INC
Substrate for plasmin and microplasmin. Microplasmin hydrolyzed the peptide substrate with a Km of 0.361 +/- 0.017 mM and a kcat of 40.3 +/- 3.3 s⁻¹, whereas native plasmin had a Km of 0.355 +/- 0.002 mM and a kcat of 27.9 +/- 0.3 s⁻¹ (50 mM Tris-HCl, pH 7.4, 0.1 M NaCl, 37 °C).