Boc-Ala-Ala-Gly-pNA
Supplier: BACHEM AMERICAS INC
4006118.0250
L-1170.0250BAEA 883.44 USD
L-1170.0250BA
Boc-Ala-Ala-Gly-pNA
Proteins and Peptides
Boc-AAG-pNA, substrate for glycine endopeptidase (chymopapain M) from Papaya carica.