H-Ala-Ala-Pro-pNA · HCl
Supplier: BACHEM AMERICAS INC
4004827.0050
L-1100.0050BAEA 735.3 USD
L-1100.0050BA
H-Ala-Ala-Pro-pNA · HCl
Proteins and Peptides
AAP-pNA, chromogenic substrate for dipeptidyl aminopeptidase yscV, a thermosensitive enzyme from yeast and for prolyl tripeptidyl peptidases.