KEMRESIN® Venturi Work Tops, Kewaunee®
Supplier: Kewaunee
VBTR012448-AC
KVBTR012448-AEA 1554.06 USD
KVBTR012448-A KVBTR012448-AC KVBTR012460-A KVBTR012460-AC KVBTR012472-A KVBTR012472-AC KVBTR012496-A KVBTR012496-AC
KEMRESIN® Venturi Work Tops, Kewaunee®
Fume Hoods and Enclosures Chemical Fume Hoods
Venturi fume hood worktop can be used wherever optimum physical and chemical resisting properties are required.
  • Ultra-smooth non-porous surface
  • Composed of 1" and ³/₄" thick molded epoxy resin
  • Resistant to all alkalis, acids and solvents
  • Highly resistant to heat and physical abuse
It also contains features such as left rear cupsink, horizontal flush.
Certifications: Meet SEFA 3 chemical resistance performance standards.
Ordering information: Available in the colors: black, putty, gray, slate.