Venturi V05-General Purpose Bench Fume Hood with Vertical Rising Sash, Kemglass Liner, Kewaunee Scientific

Supplier: Kewaunee
V05F282448GN-V
KSV05F282448GMEA 14352.5 USD
KSV05F282448GM KSV05F282448GN KSV05F282448GS KSV05F282460GM KSV05F282460GN KSV05F282460GS KSV05F282472GM KSV05F282472GN KSV05F282472GS KSV05F282496GM KSV05F282496GN KSV05F282496GS KSV05F283048GM KSV05F283048GN KSV05F283048GS KSV05F283060GM KSV05F283060GN KSV05F283060GS KSV05F283072GM KSV05F283072GN KSV05F283072GS KSV05F283096GM KSV05F283096GN KSV05F283096GS KSV05F283648GM KSV05F283648GN KSV05F283648GS KSV05F283660GM KSV05F283660GN KSV05F283660GS KSV05F283672GM KSV05F283672GN KSV05F283672GS KSV05F283696GM KSV05F283696GN KSV05F283696GS KV05F282448GNV KV05F282472GNV
Venturi V05-General Purpose Bench Fume Hood with Vertical Rising Sash, Kemglass Liner, Kewaunee Scientific
Fume Hoods and Enclosures Chemical Fume Hoods
The Supreme Air Venturi provides the safest user-friendly fume hood manufactured today. The Supreme Air Venturi utilizes proven containment enhancing techniques coupled with innovative laboratory convenience features, including: adjustable LED lights, electromechanical sash stops, ultra-smooth sash system, and push button sash management.

  • Un paralleled Venturi enhanced containment technology
  • High performance/energy efficient
  • Unique ultra-smooth sash system
  • Exceptional viewing height
  • Flush airfoil
  • LED lights with color and intensity adjustability
  • Electromechanical sash stops

Color: Light neutral or snow white

Certifications: UL and cUL listed. Tested to ASHRAE 110, NIH03-112C, ASTM D552, and NFPA 45.

Ordering information: Work top and base cabinets not included with fume hood.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR