KIMBLE® ULTRA-WARE® Conversion Base, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
953984-0090
KT953984-0090EA 985.62 USD
KT953984-0090
KIMBLE® ULTRA-WARE® Conversion Base, DWK Life Sciences
Filters Membrane Filter Units
ULTRA-WARE® conversion base converts the five valve, four valve and filtration caps to use 90 mm filter membranes, providing up to four times faster filtration.
  • Requires a 90 mm solvent pickup adapter
The conversion base is manufactured from glass-filled PTFE and is supplied with a coarse porosity (40 to 60 micron porosity) glass support frit and an upper screw clamp.