KIMBLE® ULTRA-WARE® Economy Three-Hole Cap Systems, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
953902-1002 953902-5002 953930-1002 953902-0252 953902-0502 953902-1003 953902-2002 953930-1003 953930-2002 953930-5002
KT953902-1002EA 275.43 USD
KT953902-1002 KT953902-5002 KT953930-1002 KT953902-0252 KT953902-0502 KT953902-1003 KT953902-2002 KT953902-2003 KT953930-1003 KT953930-2002 KT953930-5002
KIMBLE® ULTRA-WARE® Economy Three-Hole Cap Systems, DWK Life Sciences
Chromatography Systems Complete HPLC Systems
The ULTRA-WARE® Reservoir System with reservoir bottles are made up of Type I, Class A borosilicate glass and PTFE cap body.
The glass with a plastic safety coating blocks virtually all UV light up to 385 nm, preventing photodegradation of light sensitive mobile phases.
The connecting threads on the top of the cap use standard 1/4-28 flangeless fittings. Two of the ports have 1/8” through holes, the third port has a 1/16” through hole.
Certifications: Conforms to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery information: Economy Three Hole Cap Systems areMore Product Information