KIMBLE® ULTRA-WARE® Delivery Caps, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
953913-0000
KT953913-0000EA 348.67 USD
KT953913-0000
KIMBLE® ULTRA-WARE® Delivery Caps, DWK Life Sciences
Closures Safety Caps and Safety Waste Caps
Caps have 1/4"-28 threads top and bottom with 1/8" ID holes to accept 1/8" OD PTFE tubing
Supplied complete with three 1/4"- 28 CTFE plugs and a TFE/propylene o-ring.