KIMBLE® ULTRA-WARE® HPLC Reservoir Adapter, GL45, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
953905-0000
KT953905-0000EA 377.75 USD
KT953905-0000
KIMBLE® ULTRA-WARE® HPLC Reservoir Adapter, GL45, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
Designed to convert GL 45 threads to standard taper 40/35 inner joints.
  • Autoclavable
  • Allows filter membrane support glassware with 40/35 outer joints to be used on ULTRA-WARE®
  • Highly chemically resistant PTFE remains nonbrittle at sub-zero temperatures