KIMBLE® KONTES® ULTRA-WARE® Funnel/Support Assembly, 47 mm Stainless Steel Support, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
953805-0000
KT953805-0000EA 442.65 USD
KT953805-0000
KIMBLE® KONTES® ULTRA-WARE® Funnel/Support Assembly, 47 mm Stainless Steel Support, DWK Life Sciences
Filters Membrane Filter Units
This 47 mm vacuum filtration apparatus is designed to handle larger volumes of liquids for analysis of particulate or biological contamination.
This unit has a removable, 316 stainless screen filter support. An aluminum clamp sandwiches filter of choice between funnel and base. Connection to KT953760-0000 1 L flask (available as an accessory) is made with a silicone stopper.
Also mounts in Millipore® filtering manifolds.