KIMBLE® Fritted Discs, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
952000-8044
KT952000-8044EA 207.38 USD
KT952000-8044
KIMBLE® Fritted Discs, DWK Life Sciences
Filters Filter Discs
Individual fritted discs for the fabrication of special apparatus or as a separate filtering device.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.