KIMBLE® NMR Sealing Manifold, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
897047-0001
KT897047-0001CS 247.75 USD
KT897047-0001
KIMBLE® NMR Sealing Manifold, DWK Life Sciences
Manifolds
NMR sealing manifold provides an easy way to evacuate and tip-off NMR tubes without additional sealing to other glass apparatus.
  • Utilize Kimble® HI-VAC® valves
  • All O-rings supplied are FKM
  • O-ring size shaft - tip: 110-011
NMR manifolds feature a BEVEL-SEAL™ threaded connection and a specially designed cap with an aluminum shield for heat deflection.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.