Antibiotic Moisture Content Flasks, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
881700-0000
KT881700-0000EA 155.69 USD
KT881700-0000
Antibiotic Moisture Content Flasks, Kimble Chase, DWK Life Sciences
Flasks Distillation Flasks
For determining moisture loss on drying in tablets, troches, or capsules.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.