KONTES® Pennyhead [ST] Stoppers, Full Length, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
851000-1030 851000-2440 851000-2942 851000-3445 851000-1435 851000-1938 851000-4550
KT851000-1030EA 43.33 USD
KT851000-1030 KT851000-2440 KT851000-2942 KT851000-3445 KT851000-1435 KT851000-1938 KT851000-4550
KONTES® Pennyhead [ST] Stoppers, Full Length, Kimble Chase
Stoppers Glass Stoppers
Glass [ST] stoppers for use in the same size outer joints
Sizes [ST]10/30 through [ST]19/38 are solid. Sizes [ST]24/40 and larger are hollow.