KIMBLE® Rack for Autosampler Vials, DWK Lifesciences
Supplier: DWK Life Sciences (KIMBLE)
749210-0012
KT749210-0012PK 147 USD
KT749210-0012
KIMBLE® Rack for Autosampler Vials, DWK Lifesciences
Racks Chromatography Vial Racks
Disposable vial racks made of polypropylene.
  • Lettered and numbered for indexing individual tubes
  • Corners interlock for convenient stacking