KIMBLE® Filtering Tube, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
748021-1030
KT748021-1030CS 52.98 USD
KT748021-1030
KIMBLE® Filtering Tube, DWK Life Sciences
Filters Filter Tubes
This tube is suitable for filtrations in semi-micro analysis and is tooled for use with a KT748012-1000 Hirsch funnel.
  • With a reinforced lip
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.