KIMBLE® Plastic Luer Slip Micro Syringe, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
748019-0001
KT748019-0001CS 19.12 USD
KT748019-0001
KIMBLE® Plastic Luer Slip Micro Syringe, DWK Life Sciences
Syringes Syringes for Chromatography
The Plastic Luer Slip Micro Syringes work for chromatography, nuclear medicine, HPLC sample preparation, and many laboratory procedures.
  • Two Part
  • Plastic
  • Polypropylene Barrel
  • Polyethylene Plunger
  • Latex Free
More chemically resistant than rubber-tipped syringes, these two-part plastic syringes feature a polypropylene barrel and polyethylene, low dead-space plunger
The needles are for research use only and are not for use in diagnostic procedures.
The sterile, individually wrapped needles contain no rubber, silicone oil, styrene, DNA, latex, or DEHP.
Ordering information: Syringes are supplied without needles.