KONTES® Craig Recrystallization Tube Set, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
747330-0003
KT747330-0003EA 58.08 USD
KT747330-0003
KONTES® Craig Recrystallization Tube Set, Kimble Chase
Tubes Recrystallization Tubes
Recrystallization set consists of a Craig tube and a glass plug.
Plug is placed in the tube neck, and the entire apparatus is inverted into a test or centrifuge tube. Centrifuging the apparatus will form a mat of crystals between the plug and the walls of the Craig tube while drawing away excess solvent.