Connecting Adapter, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
747160-0000
KT747160-0000EA 33.28 USD
KT747160-0000
Connecting Adapter, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
Glassware is manufactured from 33 expansion, low extractable borosilicate glass.
  • Ideal for use to 200 °C
They can be assembled 'right side up' or 'upside down', providing great versatility.
Certifications: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.