KIMBLE® Glass Threaded Microscale Connecting Adapters with Outer Standard Tapered Joints, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
747130-1314 747130-1319 747130-1324 747130-2014 747130-2024
KT747130-1314EA 62.66 USD
KT747130-1314 KT747130-1319 KT747130-1324 KT747130-2014 KT747130-2019 KT747130-2024
KIMBLE® Glass Threaded Microscale Connecting Adapters with Outer Standard Tapered Joints, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
Connecting adapters allow interfacing of Standard Taper ground jointed glassware with threaded microscale components.
  • Connects standard taper outer ground joint to GPI thread of inner ground joint
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.