KIMBLE® PTFE Threaded Microscale Connecting Adapters with [ST] Inner Joint, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
747128-2014 747128-2019
KT747128-2014EA 66.4 USD
KT747128-2014 KT747128-2019
KIMBLE® PTFE Threaded Microscale Connecting Adapters with [ST] Inner Joint, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
Adapters allow interfacing between outer Standard Taper jointed glassware with threaded microscale components.
  • Threaded on one end
  • With [ST] inner joint
  • Manufactured from PTFE
Provides a shorter travel path than the equivalent glass connector.