Claisen Adapter, Threaded, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
747110-0000
KT747110-0000EA 93.16 USD
KT747110-0000
Claisen Adapter, Threaded, DWK Life Sciences
Distillation Components Distillation Adapters
This adapter converts single-neck flasks or vials to three-neck vessels for chemical reactions such as Grignard reactions.
  • Allows easy, vacuum-tight connections of syringes for liquid addition, thermometers or gas inlet tubes
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.