Graduated Midget Impingers, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
737550-0000
KT737550-0000EA 181.44 USD
KT737550-0000
Graduated Midget Impingers, Kimble Chase
Samplers Air Samplers
This impinger was designed for the sampling of small air volumes with a low jet velocity
Similar design to the Greenburg-Smith impinger, except for its smaller size. Requires only a 12” head of water. Orifice at the bottom of the impinger is 1 mm, passing 0.09-0.11 CFM of air at 12” H2O vacuum. Pieces are interchangeable, maintaining nozzle centering and clearance to the bottom inside surface.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.