Airless Gas Bubbler, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
652220-0000
KT652220-0000EA 108.06 USD
KT652220-0000
Airless Gas Bubbler, Kimble Chase
Bubblers
Used with mineral oil to observe gaseous addition to reactions
May be used to vent a reaction mixture to the atmosphere. Tubing connections are 8 mm O.D. and may be used with an adapter to join multiple bubblers if necessary.