Ungraduated Addition Funnels with PTFE Stopcock, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
29034F-500 633030-0250 633030-0500 633031-0250
30358-225CS 424.02 USD
30358-225 KT633030-0250 KT633030-0500 KT633031-0250 KT633031-0500
Ungraduated Addition Funnels with PTFE Stopcock, DWK Life Sciences
Funnels Dropping Funnels
Cylindrical addition funnel with PTFE stopcock. Available with or without a glass hex head stopper.
  • 24/40 [ST] bottom joint with drip
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirement.