KIMBLE® Distilling Flasks, Jacketed, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
609500-0524
KT609500-0524EA 494.65 USD
KT609500-0524
KIMBLE® Distilling Flasks, Jacketed, DWK Life Sciences
Flasks Distillation Flasks
Jacketed three-neck round-bottom reaction flask with standard taper outer joints, all vertical.
  • Heavy wall
  • Manufactured from 33 expansion, low extractable borosilicate glass
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.