KIMBLE® KONTES® Round-Bottom Boiling Flasks, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
601000-0124 601000-0224 601000-0324 601000-0424 601000-0629 601000-0724 601000-0745 601000-0824 601000-0829 601000-0929 601000-1024 601000-1045 601000-2224 601000-0229 601000-0429 601000-0624 601000-0924
KT601000-0124EA 57.94 USD
KT601000-0124 KT601000-0129 KT601000-0224 KT601000-0324 KT601000-0424 KT601000-0529 KT601000-0629 KT601000-0724 KT601000-0729 KT601000-0745 KT601000-0824 KT601000-0829 KT601000-0845 KT601000-0929 KT601000-0945 KT601000-1024 KT601000-1045 KT601000-2224 KT601000-2229 KT601000-0229 KT601000-0429 KT601000-0524 KT601000-0624 KT601000-0924
KIMBLE® KONTES® Round-Bottom Boiling Flasks, DWK Life Sciences
Flasks Round Bottom Flasks
Single neck flask with a standard taper outer joint.
  • Heavy wall
  • ASTM Method D95
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.