KIMBLE® Micro Distilling Column, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
569261-0319
KT569261-0319EA 152.62 USD
KT569261-0319
KIMBLE® Micro Distilling Column, DWK Life Sciences
Distillation Components Distillation Columns
Micro-type distilling column designed to prevent flooding while retaining residue material.
  • Manufactured from 33 expansion, low extractable borosilicate glass
Kimble® KT569261-0319 micro distilling column with three chambers and 19/22 [ST] joints, no hooks, length: 122 mm
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.