KIMBLE® Distillation Receivers, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
535801-0000
KT535801-0000EA 812.49 USD
KT535801-0000
KIMBLE® Distillation Receivers, DWK Life Sciences
Distillation Components Distillation Receivers
These distillation receivers are ideally suited for the determination of water content in organic solvents.
  • Unit is compact in both the standard taper 24/40 and standard taper 14/20 sizes
  • Lower three-way stopcock provides a sample port, and the apparatus can be drained without disassembly
  • Trap area is positioned well above the boiling flask
  • Thermometer joint is standard taper 10/30. Stopcock plug is KT822501-0002
  • Manufactured from 33 expansion, low extractable borosilicate glass
Dean Stark design for solvents that are classified as either heavier than water or lighter than water. Design suggested by Dr. A. J. East, Celanese Corp.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.