KIMBLE® Petrochemical Filter Stick Apparatus with Hooks and Springs, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
513880-0000
KT513880-0000EA 187.98 USD
KT513880-0000
KIMBLE® Petrochemical Filter Stick Apparatus with Hooks and Springs, DWK Life Sciences
Petroleum Testing Apparatus
Apparatus used in the determination of oil content and solvent extractables in petroleum waxes.
  • Ref: ASTM D721
  • Manufactured from 33 expansion, low extractable borosilicate glass
Assembly consists of a sintered glass filter stick with air pressure inlet tube and delivery nozzle and a cooling tube with 24/40 joints.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery information: Supplied with one pair of springs.