KONTES® Three-Chamber Distillation Column, Snyder, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
503000-0121
KT503000-0121EA 271.99 USD
KT503000-0121
KONTES® Three-Chamber Distillation Column, Snyder, Kimble Chase
Distillation Components Distillation Columns
Snyder distillation column with floating ball valves for improved vapor-liquid contact
Approx. overall height: 305mm. Column length (without joint): 225mm. Chamber length: 50mm.