KIMBLE® MIDI-VAP 4000 Ammonia/Phenol Distillation System Complete, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
479490-4000
KT479490-4000CS 26647.4 USD
KT479490-4000
KIMBLE® MIDI-VAP 4000 Ammonia/Phenol Distillation System Complete, DWK Life Sciences
Digestion Systems Trace Element Detection Apparatus
MIDI-VAP 4000 systems feature a pre-programmed controller with automated shut-off. The Watlow SD digital controller is pre-programmed, with easy three-step setup, automatic shut-off, easy to read LED display and optional temperature programming.
  • PTFE gaskets prevent spills
  • Heavy-duty insulated block heats rapidly and isolated wiring eliminates damage from spills
  • Easy to adjust needle valves provide accurate vacuum control
Systems use a 50 ml sample resulting in cost-savings and reduced waste disposal. The 10-position manifold heater block requires minimal lab bench space.
Temperature range: 0 to 190 ºC
Cyanide temperature 165 ºC
Ordering information: 220 V, CE unit available by quote. 1 year warranty.