KONTES® Utility Washer for Cuvette Cells and NMR Tubes, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
459960-0000
KT459960-0000EA 492.95 USD
KT459960-0000
KONTES® Utility Washer for Cuvette Cells and NMR Tubes, Kimble Chase
Glassware Washers
For rapid, safe washing and drying of cuvettes, cells, tubes and small scale volumetric items up to Approximately 22 mm O.D
Operates efficiently on a conventional sink aspirator. Flushing is activated by lightly pressing the inverted cell onto a neoprene gasket. Recommended for 10 mm NMR tubes
Ordering information: Complete unit is supplied with one gasket. Hose connection is No. 2.