KIMBLE® Coil-Type Reflux Condenser with Two Upper Hose Barbs and Angled Outer Joint, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
457050-0000
KT457050-0000EA 538.45 USD
KT457050-0000
KIMBLE® Coil-Type Reflux Condenser with Two Upper Hose Barbs and Angled Outer Joint, DWK Life Sciences
Condensers
Coil-type condenser for greater cooling capacity.
  • Hook above lower standard taper joint for use with a KT675500-3445 KEM-KLAMP™
  • Manufactured from 33 expansion, low extractable borosilicate glass
KT675000-0028, KT675000-0040 polyacetal standard taper clamps may also be used.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.