Kimble® Nylon Three-Way Stopcock with Luer Fittings, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
420163-4503
KT420163-4503PK 133.87 USD
KT420163-4503
Kimble® Nylon Three-Way Stopcock with Luer Fittings, Kimble Chase
Stopcocks
Three-way stopcock with two female Luers to one male Luer-lock
The body and plug are nylon.