Shorty TLC Developing Tank, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
416180-1020
KT416180-1020CS 341.68 USD
KT416180-1020
Shorty TLC Developing Tank, Kimble Chase
TLC Development Tanks
The standard 20 x 20 cm TLC plate tank is difficult to use with 10 x 10 cm plates, so the “SHORTY” tank was developed
Lids are available with holes drilled for #0 stoppers to allow addition of reagents without disturbing the equilibrium of the saturated environment.
It will accommodate 5 x 10 or 10 x 20 plates, turned long side down. Both have a centering baffle to support plates during development and are supplied with lids.