TLC Plate Rack, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
416175-0000
KT416175-0000EA 399.79 USD
KT416175-0000
TLC Plate Rack, Kimble Chase, DWK Life Sciences
Racks TLC Plate Racks
TLC plate racks for use with the TLC rectangular developing tanks.
  • Holds 20×20 cm plates