Quartz Beaker, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
319010-0050
KT319010-0050EA 347.85 USD
KT319010-0050
Quartz Beaker, Kimble Chase
Beakers
These Quartz beakers are designed for muffle furnace ashing procedures
Useful where chemical purity is a factor in subsequent analysis. Quartz will also satisfy high temperature requirements to 1050ºC. Very useful for determining trace metals in environmental samples.