KIMBLE® Cow Type Distillation Receiver with Three Receivers and 14/20 [ST] Joints, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
287800-0000
KT287800-0000EA 492.7 USD
KT287800-0000
KIMBLE® Cow Type Distillation Receiver with Three Receivers and 14/20 [ST] Joints, DWK Life Sciences
Distillation Components Distillation Receivers
Distillation receivers 14/20 cow type with three receivers.
  • Cow-type design having three receivers spaced 45° apart
  • Manufactured from 33 expansion, low extractable borosilicate glass
Condensers with drip joints will protrude into the spherical section, keeping wetted surfaces and product hold-up to a minimum.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.