KIMBLE® Short Path Distillation Heads, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
287110-0000
KT287110-0000CS 554.65 USD
KT287110-0000 KT513750-0000
KIMBLE® Short Path Distillation Heads, DWK Life Sciences
Distillation Components Distillation Heads
This close-coupled condenser and vacuum connection distillation head offers an extremely short condensate travel path.
  • Manufactured from 33 expansion, low extractable borosilicate glass
When a standard taper thermometer is inserted in the top of this apparatus, the bulb and stem serve as column packing.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.